Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Rabbit anti-Mouse, Polyclonal, Biologo Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$517.28 - $1064.80

Specifications

Antigen Fibronectin
Concentration 1 mg/mL
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-210-0770 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
FI24951
0.5 mL
Each for $1,064.80
Add to cart
 
50-210-0771 Non-distribution product as customer accommodation.
View Documents
Exalpha Biologicals IncSupplier Diversity Partner
FI2495101
0.1 mL
Each for $517.28
Add to cart
 
Description

Description

Fibronectin Polyclonal antibody specifically detects Fibronectin in Mouse samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays
Specifications

Specifications

Fibronectin
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Radioimmune Assays
Unconjugated
Rabbit
Mouse
P11276
14268
IgG
Primary
-20°C
FN1
1 mg/mL
Polyclonal
Lyophilized
RUO
PBS with no preservative; pH
Anastellin; CIG; Cold-insoluble globulin; cumulus cell-specific fibronectin 1 transcript; DKFZp686F10164; DKFZp686H0342; DKFZp686I1370; DKFZp686O13149; E330027I09; ED-B; embryo-specific fibronectin 1 transcript; ferritin L subunit; ferritin L-chain; FIBNEC; Fibronectin; fibronectin 1; fibronectin ED-A- region; fibronectin ED-B+ region; fibronectin variable region; FINC; Fn; FN1; fn-1; FNZ; GFND; GFND2; LETS; LOW QUALITY PROTEIN: fibronectin; migration-stimulating factor; MSF; Ugl-Y1; Ugl-Y2; Ugl-Y3
Purified fibronectin from murine plasma
Polyclonal
Affinity chromatography
Antibody
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit