Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibulin-3/EFEMP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$268.00 - $706.50
Specifications
Antigen | Fibulin-3/EFEMP1 |
---|---|
Dilution | Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100 |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018172
![]() |
Novus Biologicals
NB3006910.025MG |
0.025 mg |
Each for $268.00
|
|
|||||
NB300691005
![]() |
Novus Biologicals
NB3006910.05MG |
0.05 mg |
Each for $426.00
|
|
|||||
NB300691
![]() |
Novus Biologicals
NB300691 |
0.1 mg |
Each for $706.50
|
|
|||||
Description
Fibulin-3/EFEMP1 Polyclonal specifically detects Fibulin-3/EFEMP1 in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
Fibulin-3/EFEMP1 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, ELISA | |
Unconjugated | |
Rat, Monkey, Mouse, Human | |
2202 | |
Synthetic cyclic peptide corresponding to N terminal amino acids 25-42 of Human EFEMP1, peptide was covalently modified. Sequence: TYTQCTDGYEWDPVRQQC | |
Primary |
Western Blot 1:500, ELISA 1:10000, Immunoprecipitation 1:200, Immunohistochemistry-Paraffin 1:100 | |
Polyclonal | |
Rabbit | |
DHRD, EGF containing fibulin-like extracellular matrix protein 1, EGF-containing fibulin-like extracellular matrix protein 1, Extracellular protein S1-5, FBLN3DRAD, FBNLFLJ35535, FIBL-3, fibrillin-like, Fibrillin-like protein, Fibulin-3, MGC111353, MLVT, MTLV, S1-5 | |
EFEMP1 | |
IgG | |
The Anti EFEMP1 selective was generated against a conserved sequence near the N terminal region that is unique to EFEMP1 protein only. This detects a strong 62 kDa EFEMP1 protein in human. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title