Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibulin-3/EFEMP1 Antibody (OTI1D9), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270700AF647
Description
Fibulin-3/EFEMP1 Monoclonal specifically detects Fibulin-3/EFEMP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
Fibulin-3/EFEMP1 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
EFEMP1 | |
Full length recombinant protein of human Fibulin-3/EFEMP1 (NP_004096) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Western Blot, Flow Cytometry | |
OTI1D9 | |
Western Blot, Flow Cytometry | |
DHRD, EGF containing fibulin-like extracellular matrix protein 1, EGF-containing fibulin-like extracellular matrix protein 1, Extracellular protein S1-5, FBLN3DRAD, FBNLFLJ35535, FIBL-3, fibrillin-like, Fibrillin-like protein, Fibulin-3, MGC111353, MLVT, MTLV, S1-5 | |
Mouse | |
Affinity Purified | |
Vision | |
2202.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction