Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibulin-3/EFEMP1 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI1D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270700JF549

Catalog No. N270700J549


Add to cart

Description

Description

Fibulin-3/EFEMP1 Monoclonal antibody specifically detects Fibulin-3/EFEMP1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Fibulin-3/EFEMP1
OTI1D9
Flow Cytometry
Affinity Purified, Conjugated
EFEMP1
Full length recombinant protein of human Fibulin-3/EFEMP1 (NP_004096) produced in HEK293T cells.
Immunogen affinity purified
Vision
Primary
2202.0
Flow Cytometry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
DHRD, EGF containing fibulin-like extracellular matrix protein 1, EGF-containing fibulin-like extracellular matrix protein 1, Extracellular protein S1-5, FBLN3DRAD, FBNLFLJ35535, FIBL-3, fibrillin-like, Fibrillin-like protein, Fibulin-3, MGC111353, MLVT, MTLV, S1-5
Mouse
IgG2a
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit