Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibulin 5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550961

Catalog No. PA550961


Add to cart

Description

Description

Fibulin 5 Polyclonal Antibody detects endogenous levels of Fibulin 5 protein.

Essential for elastic fiber formation, is involved in the assembly of continuous elastin (ELN) polymer and promotes the interaction of microfibrils and ELN (PubMed:18185537). Stabilizes and organizes elastic fibers in the skin, lung and vasculature. Promotes adhesion of endothelial cells through interaction of integrins and the RGD motif. Vascular ligand for integrin receptors which may play a role in vascular development and remodeling (PubMed:10428823). {ECO:0000250; UniProtKB:Q9WVH9, ECO:0000269; PubMed:10428823, ECO:0000269; PubMed:18185537}.
Specifications

Specifications

Fibulin 5
Polyclonal
Unconjugated
FBLN5
Liquid
FBLN5
Synthesized peptide derived from the Internal region of human Fibulin-5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q9UBX5, Q9WVH9, Q9WVH8
DANCE
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10516, 23876, 29158
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit