Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibulin 7 Rabbit anti-Human, Polyclonal, Bioss
Rabbit Polyclonal Antibody
Supplier: Bioss BS13161R
Description
An adhesion molecule that interacts with extracellular matrix molecules in developing teeth and may play important roles in differentiation and maintenance of odontoblasts as well as in dentin formation.Specifications
Fibulin 7 | |
Polyclonal | |
Unconjugated | |
FBLN7 | |
FBLN7; FIBL-7; fibulin 7; Fibulin-7; TM14 | |
Rabbit | |
Protein A | |
RUO | |
129804 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q53RD9 | |
FBLN7 | |
KLH conjugated synthetic peptide derived from human Fibulin 7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction