Learn More
Ficolin-3 Mouse anti-Human, DyLight 488, Clone: FCN334, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP141283G
Catalog No. NBP141283G
Add to cart
Description
Description
Ficolin-3 Monoclonal antibody specifically detects Ficolin-3 in Human samples. It is validated for ELISA.
Specifications
Specifications
Ficolin-3
FCN334
ELISA
Purified
FCN3
H-Ficolin (Hakata antigen, Ficolin-3) is an innate immunity pattern recognition molecule found in human serum. H-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP) formed by carbohydrates (GlcNAc, GalNAc and fucose) activating complement through the lectin pathway by means of the associated MASPs ((MBL)-associated serine proteases). H-Ficolin consists of 35 kDa protein chains associated in triplet subunits which are again associated in high molecular weight oligomers by means of disulfide bonds at the N-terminal region.
Protein A purified
Store at 4C in the dark.
Monoclonal
Reacts specifically with H-ficolin. No cross-reaction has been observed with L- or M-ficolin.
ELISA
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Collagen/fibrinogen domain-containing lectin 3 p35, Collagen/fibrinogen domain-containing protein 3, FCNHficolin 3, ficolin (collagen/fibrinogen domain containing) 3 (Hakata antigen), ficolin (collagen/fibrinogen domain-containing) 3 (Hakata antigen), ficolin-3, HAKA1MGC22543, Hakata antigen, H-ficolin
Mouse
IgG1 Kappa
0.1 ml
Primary
8547
Human