Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More

Ficoll 400 (White to Off-White Powder/Molecular Biology), Fisher BioReagents™
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Poly Bottle
Description
Ficoll 400 (White to Off-White Powder/Molecular Biology)
Ficoll 400 is used as a component of Denhardt's Reagent and in gel-loading buffers for DNA gels. It is also used in forming density gradients to separate cells and subcellular components.
Specifications
Color | White |
Physical Form | Powder/Solid |
Quantity | 25 g |
Formula Weight | ≈400,000 |
Grade | Molecular Biology |
Packaging | Poly Bottle |
Specific Rotation | Inclusive between 50° (+ or -) |
Chemical Name or Material | Ficoll™ 400 |
Safety and Handling

Recommended Storage : RT
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title