Learn More
Fieldmaster™ Lake Bottom Sampling Kit
Complete kit for obtaining samples from the bottom of a pond or lake
Supplier: Science First 6237210
Includes: Plastic wash bucket for sorting, armored glass thermometer from -40/+120°F, Mighty Grab, 500mL polypropylene square wash bottles (12), set of comprehensive teacher instructions.
Description
Durable, reusable apparatus.
- Might Grab™ needs no messenger, can be operated by either hand line or included sampling pole
- Bucket has 504 micron mesh
Specifications
Obtaining samples from the bottom of a pond or lake | |
Plastic wash bucket for sorting, armored glass thermometer from -40/+120°F, Mighty Grab, 500mL polypropylene square wash bottles (12), set of comprehensive teacher instructions |