Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIG4 Mouse anti-Human, Mouse, Rat, Clone: S202-7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527648

Catalog No. PIMA527648


Add to cart

Description

Description

The PI(3,5)P2 regulatory complex regulates both the synthesis and turnover of phosphatidylinositol 3,5-bisphosphate (PtdIns(3,5)P2). In vitro, hydrolyzes all three D5-phosphorylated polyphosphoinositide substrates in the order PtdIns(4,5)P2 > PtdIns(3,5)P2 > PtdIns(3,4,5)P3. Plays a role in the biogenesis of endosome carrier vesicles (ECV)/multivesicular bodies (MVB) transport intermediates from early endosomes.
Specifications

Specifications

FIG4
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Q91WF7, Q92562
Polyphosphoinositide phosphatase, 5-bisphosphate 5-phosphatase, Fig4, KIAA0274, Sac3, Phosphatidylinositol 3, Phosphatidylinositol 3,5 bisphosphate 5 phosphatase, SAC domain containing protein 3, SAC domain-containing protein 3, SAC3
Mouse
IgG1
100 μg
-20°C
Primary
103199, 309855, 9896
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
S202-7
Unconjugated
FIG4
Liquid
Fig4
Fusion protein amino acids 688-907 (C-terminus) of mouse Sac3. Rat: 95% identity (203/220 amino acids identical). Human: 91% identity (201/220 amino acids identical)
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit