Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FIG4 Mouse anti-Human, Mouse, Rat, FITC, Clone: S202-7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242197F

Catalog No. NBP242197F


Add to cart

Description

Description

FIG4 Monoclonal antibody specifically detects FIG4 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

FIG4
S202-7
Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
FIG4
Fusion protein amino acids 688-907 (C-terminus) of mouse Sac3. Rat: 95% identity (203/220 amino acids identical). Human: 91% identity (201/220 amino acids identical)
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects approx 100kDa.
Immunohistochemistry, Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
ALS11polyphosphoinositide phosphatase, CMT4J, dJ249I4.1, EC 3.1.3, EC 3.1.3.-, EC 3.1.3.36, FIG4 homolog (S. cerevisiae), FIG4 homolog, SAC1 lipid phosphatase domain containing (S. cerevisiae), hSac3, KIAA0274FIG4 homolog, SAC domain containing lipid phosphatase, Phosphatidylinositol 3,5-bisphosphate 5-phosphatase, Sac domain-containing inositol phosphatase 3, SAC domain-containing protein 3, SAC3FIG4 homolog, SAC1 domain containing lipid phosphatase
Mouse
IgG1
0.1 ml
Primary
9896
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit