Learn More
FIG4 Mouse anti-Human, Mouse, Rat, HRP, Clone: S202-7, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242197H
Catalog No. NBP242197H
Add to cart
Description
Description
FIG4 Monoclonal antibody specifically detects FIG4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
FIG4
S202-7
Western Blot, Immunohistochemistry
Purified
FIG4
Fusion protein amino acids 688-907 (C-terminus) of mouse Sac3. Rat: 95% identity (203/220 amino acids identical). Human: 91% identity (201/220 amino acids identical)
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects approx 100kDa.
Western Blot, Immunohistochemistry
HRP
PBS with No Preservative
ALS11polyphosphoinositide phosphatase, CMT4J, dJ249I4.1, EC 3.1.3, EC 3.1.3.-, EC 3.1.3.36, FIG4 homolog (S. cerevisiae), FIG4 homolog, SAC1 lipid phosphatase domain containing (S. cerevisiae), hSac3, KIAA0274FIG4 homolog, SAC domain containing lipid phosphatase, Phosphatidylinositol 3,5-bisphosphate 5-phosphatase, Sac domain-containing inositol phosphatase 3, SAC domain-containing protein 3, SAC3FIG4 homolog, SAC1 domain containing lipid phosphatase
Mouse
IgG1
0.1 ml
Primary
9896
Human, Mouse, Rat