Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Filaggrin Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Filaggrin Ab-1, Mouse Monoclonal Antibody.

$182.00 - $909.00

Specifications

Antigen Filaggrin Ab-1
Clone FLG01
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 7
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS449P
View Documents
Thermo Scientific™
MS449P
Purified Ab with BSA and Azide 1mL
Each for $909.00
Add to cart
 
MS449P0
View Documents
Thermo Scientific™
MS449P0
Purified Ab with BSA and Azide 100μL
Each for $182.00
Add to cart
 
MS449P1
View Documents
Thermo Scientific™
MS449P1
Purified Ab with BSA and Azide 500μL
Each for $629.50
Add to cart
 
MS449P1ABX
View Documents
Thermo Scientific™
MS449P1ABX
Ab without BSA/Azide (Frozen) 100μL
Each for $536.00
Add to cart
 
MS449PABX
View Documents
Thermo Scientific™
MS449PABX
Ab without BSA/Azide (Frozen) 200μL
Each for $797.00
Add to cart
 
MS449R7
View Documents
Thermo Scientific™
MS449R7
Ready-to-use for IHC (UVLP) 7mL
Each for $417.00
Add to cart
 
MS449R1
View Documents
Thermo Scientific™
MS449R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with Thermo Scientific™ Filaggrin Ab-1, Mouse Monoclonal Antibody.

Filaggrin is expressed only in well-differentiated, keratinized epithelial cells. The synthesis of filaggrin varies in types of ichthyosis, and with viral, premalignant and malignant conditions.

Host Species: Mouse

Clone: FLG01

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant filaggrin protein

Molecular Weight: 38 kDa (Filaggrin) and 400 kDa (Profilaggrin)

Positive Control: Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Filaggrin Ab-1
Immunohistochemistry (Paraffin)
Unconjugated
RUO
Primary
IgG2a kappa
FLG01
Monoclonal
Murine
Cytoskeletal Protein
Recombinant filaggrin protein
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit