Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Filaggrin Mouse anti-Human, Clone: rFLG/1561, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30737710UG

 View more versions of this product

Catalog No. NB122227


Add to cart

Description

Description

Filaggrin Monoclonal antibody specifically detects Filaggrin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Filaggrin
rFLG/1561
Unconjugated
10 mM PBS with 0.05% BSA
ATOD2, epidermal filaggrin, filaggrin, Profilaggrin
Recombinant human Filaggrin protein fragment (aa 198-288) (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
Human
Immunohistochemistry (Paraffin)
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 ug/ml
Purified
Mouse
IgG1 Kappa
100 μg
Cell Biology, Cellular Markers
Primary
2312
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit