Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Filaggrin Mouse anti-Human, Clone: rFLG/1561, Novus Biologicals™

Mouse Monoclonal Antibody

Specifications

Antigen Filaggrin
Clone rFLG/1561
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122227
View Documents
Novus Biologicals
NBP30737710UG
100 μg N/A
NB122228
View Documents
Novus Biologicals
NBP30737720UG
20 μg N/A
Description

Description

Filaggrin Monoclonal antibody specifically detects Filaggrin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Filaggrin
0.2 mg/ml
Immunohistochemistry (Paraffin)
Purified
RUO
Human
ATOD2, epidermal filaggrin, filaggrin, Profilaggrin
Recombinant human Filaggrin protein fragment (aa 198-288) (exact sequence is proprietary)
Monoclonal
Protein A or G purified
rFLG/1561
Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Cell Biology, Cellular Markers
10 mM PBS with 0.05% BSA
2312
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit