Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Filamin A Recombinant Rabbit Monoclonal Antibody (1B2)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA549798
Description
Filamin A Recombinant Monoclonal Antibody for IHC (P), ELISA
Filamin functions as a crosslinking protein forming a flexible link between two actin filaments. It is composed of two identical polypeptide chains each joined to the other at one end, with an actin binding site at the other. It is present in human platelets, lymphocytes, fibroblasts and smooth muscle actin.
Specifications
Filamin A | |
Recombinant Monoclonal | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P21333 | |
Flna | |
A synthesized peptide derived from human Filamin A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin) | |
1B2 | |
Unconjugated | |
Flna | |
450 kDa protein; ABP-280; ABPX; actin binding protein 280; actin-binding protein 280; alpha filamin; alpha-filamin; CSBS; CVD1; dilated pupils 2; Dilp2; Endothelial actin-binding protein; Endothelial actin-binding protein 280); F730004A14Rik; filamin; filamin 1; filamin A; filamin A, alpha; filamin, alpha; Filamin1; filamin-1; FilaminA; Filamin-A; FLN; FLN1; Flna; FLN-A; FMD; GENA 379; gizzard filamin; MNS; NHBP; Non-muscle filamin; OPD; OPD1; OPD2; RGD1560614; XLVD; XMVD | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2316 | |
-20°C or -80°C if preferred | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction