Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Filamin Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and ELISA experiments with Thermo Scientific™ Filamin Ab-1, Mouse Monoclonal Antibody.

Specifications

Antigen Filamin Ab-1
Clone FLMN01; same as PM6/317
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 1
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1211P1
View Documents
Thermo Scientific™
MS1211P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting and ELISA experiments with Thermo Scientific™ Filamin Ab-1, Mouse Monoclonal Antibody.

Filamin functions as a crosslinking protein forming a flexible link between two actin filaments. It is composed of two identical polypeptide chains each joined to the other at one end, with an actin binding site at the other. It is present in human platelets, lymphocytes, fibroblasts and smooth muscle actin.

Host Species: Mouse

Clone: FLMN01; same as PM6/317

Isotype: IgG1

Species Reactivity: Human, Mouse, Rabbit, Guinea Pig, Chicken and Rat. Others not known.

Epitope: Not determined

Immunogen: Platelet filamin

Molecular Weight: 250 kDa

Positive Control: NIH3T3 cells, Skin

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • ELISA

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

Filamin Ab-1
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RUO
Primary
IgG1
FLMN01; same as PM6/317
Monoclonal
Murine
Cytoskeletal Protein
Platelet filamin
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit