Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Filensin Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI2B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270249AF647

Catalog No. N270249A647


Add to cart

Description

Description

Filensin Monoclonal antibody specifically detects Filensin in Human samples. It is validated for Western Blot.
Specifications

Specifications

Filensin
OTI2B2
Western Blot
Affinity Purified, Conjugated
BFSP1
Recombinant protein fragment corresponding to amino acids 405-665 of human Filensin (NP_001186) produced in E.coli.
Immunogen affinity purified
Vision
Primary
631.0
Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Beaded filament structural protein 1, beaded filament structural protein 1, filensin, CP115Lens intermediate filament-like heavy, filensin, Lens fiber cell beaded-filament structural protein CP 115, LIFL-H115 KD
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit