Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Filensin Antibody (OTI2B2), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270249H
Description
Filensin Monoclonal specifically detects Filensin in Human samples. It is validated for Western Blot.Specifications
Filensin | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
BFSP1 | |
Recombinant protein fragment corresponding to amino acids 405-665 of human Filensin (NP_001186) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI2B2 | |
Western Blot | |
Beaded filament structural protein 1, beaded filament structural protein 1, filensin, CP115Lens intermediate filament-like heavy, filensin, Lens fiber cell beaded-filament structural protein CP 115, LIFL-H115 KD | |
Mouse | |
Affinity Purified | |
Vision | |
631.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction