Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Filensin Mouse anti-Human, Clone: OTI2B2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526645

Catalog No. PIMA526645


Add to cart

Description

Description

More than 99% of the vertebrate ocular lens is comprised of terminally differentiated lens fiber cells. Two lens-specific intermediate filament-like proteins, CP49 (also known as phakinin) and the protein product of this gene, filensin, are expressed only after fiber cell differentiation has begun. Both proteins are found in a structurally unique cytoskeletal element that is referred to as the beaded filament (BF). Mutations in this gene are the cause of autosomal recessive cortical juvenile-onset cataract. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

Filensin
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q12934
CP115, CP94, LIFL-H, cytoskeletal protein, 115 KD, filensin, lens fiber cell beaded-filament structural protein CP 115, lens intermediate filament-like heavy
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
631
Immunohistochemistry (Paraffin), Western Blot
OTI2B2
Unconjugated
BFSP1
Liquid
BFSP1
Human recombit protein fragment corresponding to amino acids 405-665 of BFSP1 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit