Learn More
BrandTech™ Filling Valves and Valve Mounting Tools for Bottletop Dispensers
Filling valves and tools are available for the proper cleaning and maintenance of BRAND™ bottletop dispensers.
Supplier: BrandTech™ 6739
Description
- Filling valves for the Dispensette™ III, Organic, TA and seripettor™ pro are provided with a sealing washer. The updated design of the Dispensette™ S series of bottletop dispensers has eliminated the need for a sealing washer.
- Filling valve with olive-shaped nozzle is recommended for Dispensette™ III and Organic bottletop instruments that are frequently autoclaved.
- seripettor™ valve set includes both the filling and discharge valves for the instrument.
- Calibrating-mounting tool is used for calibrating analog-adjustable Dispensette™ models, as well as removing the valves for routine cleaning or replacement.
Specifications
Autoclavable | |
ETFE, Ceramic | |
Filling valve |
Translucent/White | |
Dispensette™ S Trace Analysis | |
Non-sterile |