Learn More
Filter aid, Celite|r 545, treated with sodium carbonate, flux calcined, Thermo Scientific Chemicals
Description
Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Celite filter aids maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization. A medium to fine product for the filtration of most process streams including beer, cider, wines, water, low viscosity glues and organic chemicals and pharmaceuticals. for water purification.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong acids.
Specifications
Quantity | 1000 g |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Filter aid, Celite™ 545, treated with sodium carbonate, flux calcined |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H373-H319-H335 May cause damage to organs through prolonged or repeated exposure. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement P260-P261-P280-P305+P351+P338-P405-P501a Do not breathe dust/fume/gas/mist/vapors/spray. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
EINECSNumber | 272-489-0 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.