Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Filter aid, Celite Hyflo Super-cel™

Click to view available options
Quantity:
500g
2500g
61790-53-2
O2Si
60.083
MFCD00132803
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
dioxosilane
O=[Si]=O
61790-53-2
O2Si
60.083
MFCD00132803
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
dioxosilane
O=[Si]=O
Applications
Filter aid, Celite Hyflo Super-cel« is used as a filtration agent, a functional filler in paints, plastics, paper, rubber, adhesives, catalysts, agricultural chemicals, pharmaceuticals and other chemicals, and a mild abrasive in polishes and cleaners. It is also used in clarifying liquids and oils, as an efficient thermal insulator; in manufacture of heat insulators, fire brick, and fire- and acid-proof packing materials; as an absorbent; in absorbent dynamite; as a solid carrier in chromatography; pozzolanic admixture in concrete mixes.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with acids.
Quantity 2500g
Odor Odorless
Merck Index 14,4973
Chemical Name or Material Filter aid, Celite Hyflo Super-cel™

GHS H Statement
H350-H373-H319-H335
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : VV7311000

TSCA : TSCA