Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Filter aid, Celite Standard Super-cel, Thermo Scientific Chemicals

Catalog No. AAB219830E
Click to view available options
Quantity:
500 g
2500 g

Filter aid, Celite Standard Super-cel is used to maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Filter aid, Celite Standard Super-cel is used to maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization.

Solubility
Insoluble in water.

Notes
Incompatible with acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula O2Si
Molecular Weight (g/mol) 60.083
MDL Number MFCD00132803
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Quantity 2500 g
Odor Odorless
CAS 91053-39-3
Merck Index 14,4973
Solubility Information Insoluble in water.
Chemical Name or Material Filter aid, Celite Standard Super-cel
Hazard Category H319-H335-H373
Hazard Statement GHS H Statement
H350-H373-H319-H335
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c
EINECSNumber 319-127-6
RTECSNumber VV7311000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.