Learn More
AVANTOR MASTERFLEX (VWR LLC) 255003-03 Polypropylene absolute-rated high-efficiency filter bags; 10""L, 5 m
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0151644
Encompass_Preferred
Choose from standard felt bags, high-efficiency bags, or nylon mesh bags. Each bag measures 4" diameter. The 10"L bags have a surface area of 0.75 ft2; 18-¿"L bags have a surface area of 1.75 ft2.
Catalog No. 50-632-123