Learn More
Cole-Parmer Cole-Parmer 255039-03 Nylon monofilament filter bag; 18-1/2"L, 800 µm  DFS Item
Manufacturer:  Cole-Parmer 0151916
Choose from standard felt bags, high-efficiency bags, or nylon mesh bags. Each bag measures 4" diameter. The 10"L bags have a surface area of 0.75 ft2; 18-½"L bags have a surface area of 1.75 ft2.
Catalog No. 50-632-142