Learn More
Cole-Parmer Cole-Parmer 255029-03 Polypropylene absolute-rated high-efficiency filter bags; 18-1/2"L, 50 µm
Manufacturer: Cole-Parmer 0151690
Choose from standard felt bags, high-efficiency bags, or nylon mesh bags. Each bag measures 4" diameter. The 10"L bags have a surface area of 0.75 ft2; 18-½"L bags have a surface area of 1.75 ft2.