Learn More
Cole-Parmer Cole-Parmer KO50HE2 Size 2 high-efficiency PP absolute-rated filter bag; 200F (93C) max, 50 um pore  DFS Item
Manufacturer:  Cole-Parmer 0153170
Designed for 98% removal efficiency at the rated pore size, these bags offer three to four times the dirt-holding capacity of standard bags. In addition, they are FDA-compliant with respect to all regulations governing food-grade materials. These polypropylene absolute-rated filter bags are rated for a temperature of 200°F (93°C) maximum.
Catalog No. 50-632-214