Learn More
Cole-Parmer Cole-Parmer 255134-03 Size 2 High-Efficiency Polypropylene Absolute-Rated Filter Bag; 200°F (93°C) Maximum  DFS Item
Manufacturer:  Cole-Parmer 0153174
Designed for 98% removal efficiency at the rated pore size, these bags offer three to four times the dirt-holding capacity of standard bags. In addition, they are FDA-compliant with respect to all regulations governing food-grade materials. These polypropylene absolute-rated filter bags are rated for a temperature of 200°F (93°C) maximum.
Catalog No. 50-632-215