Learn More
Cole-Parmer Cole-Parmer 155162-43 Double open end long-life carbon block cartridge; 0.5 µm, 10"L  DFS Item
Manufacturer:  Cole-Parmer 0150893
Long-Life Carbon Block Cartridges have a polyolefin-bonded powdered activated carbon (PAC) filter. Surface area of 2.2 million sq ft. Removes tastes, odors, chlorine, and sediment. Contains approximately 12 oz of carbon. PP end caps and outer wraps. Maximum temperature is 100°F (38°C); maximum differential pressure is 150 psid. For optimal performance, change cartridges at 25 psid.
Catalog No. 50-632-057