Learn More
Cole-Parmer Cole-Parmer WP5BB97P Fibrous polypropylene string-wound cartridge, 5 µm, 10" length  DFS Item
Manufacturer:  Cole-Parmer 0150876
Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165°F (74 °C).
Catalog No. 50-632-263