Learn More
Cole-Parmer Double Open End Carbon-Impregnated Paper Cartridge; 5um, 10"L, 1/Pk  DFS Item
Manufacturer:  Cole-Parmer 0150925
This cellulose cartridge removes organic tastes/odors and chlorine, plus reduces sediment. Suitable to 125°>F (52°>C).
Catalog No. 50-632-048