Learn More
AVANTOR MASTERFLEX (VWR LLC) Big Blue Filter cartridge, 10"", pleated cellulose, 20 m
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0151274
Encompass_Preferred
Pleated cartridges separate viscous sediments and flat particulates. Use polyester cartridges for water filtration and polypropylene cartridges with harsher chemicals. Plastisol end caps.
Catalog No. 50-632-088