Learn More
Agilent BioTek Filter Cube
For use with Synergy LX
$1200.15
Catalog Number | Mfr. No. | Excitation/Emission | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Excitation/Emission | Price | Quantity & Availability | |||||
BT1505006
|
Agilent BioTek
1505006 |
EX 360/40, EM 460/40, 400 nm mirror (Blue) |
Each for $1,200.15
|
|
BT1505005
|
Agilent BioTek
1505005 |
EX 485/20, EM 528/20, 510 nm mirror (Green) |
Each for $1,200.15
|
|
BT1505004
|
Agilent BioTek
1505004 |
EX 530/25, EM 590/25, 570 nm mirror (Red) |
Each for $1,200.15
|
|
Specifications
Synergy LX |