Learn More
Chemglass Life Sciences Filter Funnel, Disposable, Cone Shaped, 8mL, Polypropylene, Polyethylene Frit
SDP
Supplier:  Chemglass Life Sciences OP660508
Encompass_Preferred
Filter Funnel, Disposable, Cone Shaped, 8mL, Polypropylene, Polyethylene Frit. Disposable filter funnel for solid-liquid separation can be used in chemistry and biological laboratories. The molded funnel has a 10 micron polyethylene fritted disc. Polypropylene filter funnel is designed for use with the CG-1049-J style glass vacuum adapter and a flask or vial. The CG-1049-J vacuum adapters are specially designed so that the vacuum can be controlled using a PTFE stopcock to prevent the filtrates from being sucked into the vacuum line. Also, the disposable filter funnels have an 80mm long stem so that the glass vacuum adapter is not contaminated during the filtration process. U.S. Patent Pending.
Catalog No. 50-131-1413
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.