Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva Filter Holder
Polysulfone In-Line Filter Holder for 50mm Diameter Membrane
Supplier: Cytiva 10464100
Description
Polysulfone In-Line Filter Holder for 50mm Diameter Membrane
Specifications
ML050/0 stainless steel | |
Stainless Steel | |
1 | |
50 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title