Learn More
Mettler Toledo™ Filter Kit for XPR Microbalances
Catalog No. 01911582 Shop All METTLER TOLEDO Products
Change view
Click to view available options
:
Filter Kit: Suitable for 47 and 70 mm filters, includes tweezers
Catalog No.
01-911-582 Filter Kit: Suitable for 47 and 70 mm filters, includes tweezers
1 options
Catalog No. 01-911-582 Supplier Mettler Toledo™ Supplier No. 30300922
Only null left
Add to Cart
Add to Cart
Filter weighing kit to use on XPR micro and ultra micro balances, includes tweezers suitable for filter of 47 to 70 mm diameter
  • Converts the XPR microbalance into a dedicated manual filter weighing system
  • Suitable for 47 and 70mm filters
  • Includes tweezers
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) XPR Microbalance and Ultra-Microbalance
Includes Tweezers