Learn More
Hach Company Filter, Membrane, Pore Size 0.45 µm, Diameter 47 mm, 100/pk, non-sterile, with grid
Supplier:  Hach Company 2618901
Encompass_Preferred
These 47-mm diameter, 0.45-µm pore size GN-Metricel membrane filters meet or surpass all U.S. Environmental Protection Agency (USEPA) and American Society for Testing and Materials (ASTM) specifications to ensure complete retention and maximum recovery of bacteria. Black membranes for yeast and mold are also available. Black membranes make yeast and mold colonies more distinct and easier to count. Filters are available in convenient, presterilized, individually sealed packs (P/N 13530-01 and 24903-00) or in resealable envelopes with 10 filters and 10 absorbent pads each (P/N 21269-00). Phosphate free.
Catalog No. 50-199-6370
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.