Learn More
United Scientific™ Filter Micropipette Tips, Bulk
SDPSCI_ED
Filter Micropipette Tips, Bulk are disposable tips intended for use with most brands of pipettors.
Manufacturer:  United Scientific™ UNP11005
 View more versions of this product
Catalog No. S8538023
Add to cart
Description
Description
  • Manufactured from 100% pure porous polyethylene, filters are designed to prevent liquids and aerosols from entering the body of the micropipette to protect against cross-contamination.
  • Best suited for biological applications, these tips are non-cytotoxic, non-hemolytic, and have a 99.9% bacterial filtration efficiency (BFE).
  • Tips are manufactured from ultra pure USP Class VI certified, virgin polypropylene.
  • Free of DNase, RNase, and pyrogen
Specifications
Specifications
200 μL
Non-sterile
Autoclavable
Universal
Beveled
Filtered
Natural
UNP11005
Bulk, Bagged
Yes