Learn More
Microsolv Technology CorporationSupplier Diversity Partner FILTER SF.45 CA 25MM ORANGE 10 DFS Item
Manufacturer:  Microsolv Technology CorporationSupplier Diversity Partner 58045CA25
Syringe Filters, HPLC, 25mm polypropylene housing, 0.45um Cellulose Acetate membrane. Color Code: orange. Supplied with certificate of analysis. 100/pack.
Catalog No. 50-014-52593