Learn More
Microsolv Technology CorporationSupplier Diversity Partner FILTER STONE .68 WIDE DFS Item
Manufacturer:  Microsolv Technology CorporationSupplier Diversity Partner 4822204
Filter / Sparging Stone , HPLC mobile phase inlet, stainless steel, 2um pore size, .68 in. (17.27mm) diameter OD, 1 in. (25.4mm) long, use with 1/8th in. low pressure tubing. 1 each.
Catalog No. 50-014-52503