Learn More
Thomas ScientificSupplier Diversity Partner 500mL Filter System, PES Filter Material, 0.22æm, 75mm, Sterile DFS Item
Manufacturer:  Thomas ScientificSupplier Diversity Partner 1226S78
CELLTREAT Vacuum Filter Systems are ideal for large volume sample separation and purification. The filter systems are manufactured in a Class 100,000 clean room, from USP class VI virgin polystyrene. The filter membrane is Polyethersulfone (PES) material in 0.22µm or 0.45µm pore sizes for rapid flow rate and low protein binding. The filter units are available in 150ml, 250ml and 500ml sizes and are individually packaged in easy to open plastic bags with individually wrapped caps. CELLTREAT Scientific Products include a comprehensive "Quality Certificate" with every case of product. The CELLTREAT "Quality Certificate" includes the Part Number, Lot Number, Sterility Assurance Level and Dosage, Endotoxin/Pyrogen Free Assurance, Non-Cytotoxicity Assurance and RNase/DNase Free Assurance. Filter Systems are supplied sterile (gamma irradiated). · NEW – Polyethersulfone (PES) 0.1 µm membrane is ideal for mycoplasma removal
Catalog No. NC0054838