Learn More
Basix™ Filter Tips
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
$78.00 - $210.00
Specifications
Filtered | Filtered |
---|---|
Sterility | Sterile |
Color | Natural |
Autoclavable | Not Autoclavable |
Clean Claims | DNase, RNase, DNA, and Pyrogen Free |
Catalog Number | Length (English) | Format | Volume (Metric) | Autoclavable | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Length (English) | Format | Volume (Metric) | Autoclavable | Price | Quantity & Availability | |||
13-611-104
|
1.25 in. | Rack | 10 μL | Not Autoclavable |
Pack of 960 for $111.00
Case of 5 PK for $505.00 |
![]() |
|||
13-611-105
|
1.69 in. | Rack | 10 μL | Not Autoclavable |
Pack of 960 for $107.00
Case of 5 PK for $485.00 |
|
|||
13-611-110
|
2 in. | Rack | 30 μL | Not Autoclavable |
Pack of 960 for $78.00
Case of 5 PK for $390.00 |
![]() |
|||
13-611-109
|
2 in. | Rack | 100 μL | Not Autoclavable |
Pack of 960 for $120.00
Case of 5 PK for $542.00 |
![]() |
|||
13-611-113
|
2 in. | Rack | 200 μL | Not Autoclavable |
Pack of 960 for $103.00
Case of 5 PK for $465.00 |
|
|||
13-611-114
|
3.52 in. | Rack | 200 μL | Not Autoclavable |
Pack of 1632 for $210.00
Case of 3 PK for $619.00 |
|
|||
13-611-122
|
2.312 in. | Rack | 300 μL | Not Autoclavable |
Pack of 960 for $89.00
Case of 5 PK for $404.00 |
|
|||
13-611-125
|
3.125 in. | Rack | 1000 μL | Not Autoclavable |
Pack of 1000 for $104.00
Case of 5 PK for $514.00 |
|
|||
Description
The Basix™ pipette tip portfolio consists of the most commonly used tip volumes and packaging options in todays laboratory.
Basix offers reliability and quality for all varieties of experiments, from everday liquid transfer to PCR and Electrophoresis, Basix offers a tip to meet your needs.
Choose from non-Sterile or Sterile with packaging options including bulk, racked, reloads, stack racks.
- Unique small micron filter prevents aerosol contamination with no obstruction to airflow
- Use aerosol barrier tips to ensure result integrity
- Certified by independent laboratory sterile;
- RNase, DNase, DNA, and pyrogen free
- Natural color
Specifications
Filtered | |
Natural | |
DNase, RNase, DNA, and Pyrogen Free | |
Universal | |
Basix™ |
Sterile | |
Not Autoclavable | |
Rack | |
Universal Fit Pipettor |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.