Learn More
Cole-Parmer Big Blue Filter cartridge, 10", granular carbon, 20 micron
Manufacturer: Cole-Parmer 0151264
These cartridges remove organic tastes/odors and chlorine. Special design maximizes contact time and reduces channeling or bypass that is common with GAC cartridges. Included within the cartridge is a 20-micron post-filter to reduce carbon fines††.
The 20" cartridge meets NSF Standard 42 for materials.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.