Learn More
Cole-Parmer Big Blue Filter cartridge, 10", pleated carbon, 10 micron  DFS Item
Manufacturer:  Cole-Parmer 2983032
The pleated design offers higher flow rates and dirt-loading capacities than shorter-life block cartridges. The non-cellulose media resists chemical attack and biofilm build-up making the filter ideal for both chlorinated and non-chlorinated water treatment applications. Common applications include post reverse osmosis polishing and well water treatment.
Catalog No. 50-632-087