Learn More
Sklar InstrumentsSupplier Diversity Partner Fine Mesh Basket with Locking Clip, Sklar, 8 in. x 4 in. x 1-1/2 in. Fine Mesh Basket with Locking Clip 1.6mm Openings, 0.7mm Wire Diameter, 200mm x 100mm x 35mm. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106626
Fine Mesh Basket with Locking Clip, Sklar, 8 in. x 4 in. x 1-1/2 in. Fine Mesh Basket with Locking Clip 1.6mm Openings, 0.7mm Wire Diameter, 200mm x 100mm x 35mm. Sklar's Fine Mesh Basket is designed for use with cleaning, sterilizing, and handling small instruments. The baskets are available in a variety of sizes, with or without removable hinges and with or without locks.
Catalog No. 50-143-7259