Learn More
Sklar InstrumentsSupplier Diversity Partner Fine Mesh Basket with Locking Clip, Sklar, 9-1/2 in. x 5 in. x 1-1/2 in. Fine Mesh Basket with Locking Clip 1.6mm Openings, 0.7mm Wire Diameter, 240mm x 120mm x 35mm. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 106634
Fine Mesh Basket with Locking Clip, Sklar, 9-1/2 in. x 5 in. x 1-1/2 in. Fine Mesh Basket with Locking Clip 1.6mm Openings, 0.7mm Wire Diameter, 240mm x 120mm x 35mm. Sklar's Fine Mesh Basket is designed for use with cleaning, sterilizing, and handling small instruments. The baskets are available in a variety of sizes, with or without removable hinges and with or without locks.
Catalog No. 50-143-7263