Thermo Scientific™ Finnpipette™ F1 Fixed Volume Single-Channel Pipettes

Protect samples from contamination with this manual pipette, featuring an adjustable finger rest and antimicrobial surface.

Manufacturer: Thermo Scientific™ 4651010

 View more versions of this product

Catalog No. 14-386-324

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Model Finnpipette™ F1
    Volume (Metric) 5μL
    Number of Channels One
    Accuracy +/-1.4%
    Description F1 Fixed Volume Single Channel
    Precision 1.4%

    Protect samples from contamination with the Thermo Scientific™ Finnpipette™ F1 Fixed-Volume Pipette, featuring an adjustable finger rest, antimicrobial surface and super blow-out piston with volumes of 50µL and below. The Finnpipette F1 Fixed-Volume Pipette delivers light, smooth pipetting action with patented soft-touch tip ejection for maximum user comfort. The antimicrobial surface helps keep the pipette clean without the use of additional surface decontamination liquids. The entire pipette is resistant to UV light, reagents and moisture.

    Features:

    • Antimicrobial surface: Silver (Ag) ions create a pipette surface that inhibits the growth of bacteria, microbes and fungus
    • Adjustable finger rest: Finger rest can be adjusted 120° for optimal pipetting position
    • Light, smooth pipetting action: Enables better results over longer pipetting periods
    • Super blow-out piston with volumes of 50µL and below: Ensures delivery of micro-size drops
    • Proprietary soft-touch tip ejection: Minimizes forces needed for tip ejection

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit