Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Finnpipette™ F1 Fixed Volume Single-Channel Pipettes

Protect samples from contamination with this manual pipette, featuring an adjustable finger rest and antimicrobial surface.

Manufacturer:  Thermo Scientific™ 4651040

 View more versions of this product

Catalog No. 14-386-327

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »

Disclaimers:

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


Specifications

Description

Protect samples from contamination with the Thermo Scientific™ Finnpipette™ F1 Fixed-Volume Pipette, featuring an adjustable finger rest, antimicrobial surface and super blow-out piston with volumes of 50µL and below. The Finnpipette F1 Fixed-Volume Pipette delivers light, smooth pipetting action with patented soft-touch tip ejection for maximum user comfort. The antimicrobial surface helps keep the pipette clean without the use of additional surface decontamination liquids. The entire pipette is resistant to UV light, reagents and moisture.

Features:
  • Antimicrobial surface: Silver (Ag) ions create a pipette surface that inhibits the growth of bacteria, microbes and fungus
  • Adjustable finger rest: Finger rest can be adjusted 120° for optimal pipetting position
  • Light, smooth pipetting action: Enables better results over longer pipetting periods
  • Super blow-out piston with volumes of 50µL and below: Ensures delivery of micro-size drops
  • Proprietary soft-touch tip ejection: Minimizes forces needed for tip ejection
Description & Specifications

Specifications

+/-0.6%
0.4%
1
F1 Fixed Volume Single Channel
Finnpipette™ F1
50μL
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit