Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Finnpipette™ F1 GLP Kits

Access everything needed for pipetting in these all-inclusive kits featuring state-of-the-art pipetters that maximize reliability and comfort.

Manufacturer:  Thermo Scientific™ 4700860

 View more versions of this product

Catalog No. 05-719-508

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »

Disclaimers:

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


Specifications

Description

Access everything needed for pipetting in these all-inclusive kits. Thermo Scientific™ Finnpipette™ F1 GLP (Good Laboratory Pipetting) Kit includes state-of-the-art Finnpipette F1 pipetters which offer superior performance and unique features such as adjustable finger rest and antimicrobial surface for user comfort.

Features:

  • Adjustable finger rest for increased ergonomicss and comfortability
  • Set and forget pipetting button for easy and secured volume adjustment
  • Antimicrobial surface for better protection
  • Large easy to read display for better vision ergonomics
  • Soft-touch tip ejection for light end effortless tip ejection

Kit 1 includes:

  • Three Thermo Scientific Finnpipette Pipetters from 1 to 1000µL:
  • Finnpipette F1, 1-10µL
  • Finnpipette F1, 10-100µL
  • Finnpipette F1, 100-1000µL
  • Finntip Flex tips: 10, 1x96; 200, 1x96; 1000, 1x96
  • F-stand; F1 pen; F1 brochure
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 2 includes:

  • Four Thermo Scientific Finnpipette Pipetters from 0.2 to 1000µL:
  • Finnpipette F1, 0.2-2µL
  • Finnpipette F1, 2-20µL univ
  • Finnpipette F1, 20-200µL
  • Finnpipette F1, 100-1000µL
  • Finntip Flex tips: 10, 1x96; 200, 2x96; 1000, 1x96
  • F-stand; F1 pen; F1 brochure
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 3 includes:

  • Three Thermo Scientific Finnpipette Pipetters from 10 to 10000µL:
  • Finnpipette F1, 10-100µL
  • Finnpipette F1, 100-1000µL
  • Finnpipette F1, 1000-10000µL
  • Finntip Flex tips: 200, 1x96; 1000, 1x96; FT 10mL, 1x24 F-stand; F1 pen; F1 brochure
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Kit 4 includes:

  • Three Thermo Scientific Finnpipette Pipetters from 10 to 1000µL:
  • Finnpipette F1, 2-20µL
  • Finnpipette F1, 20-200µL
  • Finnpipette F1, 100-1000µL
  • Flex tips: (2) 200µL, 10x96; 100-1000µL, 10x96
  • F-stand; F1 pen; F1 brochure
  • Good Laboratory Pipetting Guide
  • Reagent reservoir demo pack

Ordering Information:

Good Laboratory Pipetting kit also includes samples of Finntip Flex for maximum performance and best ergonomics. The kit contains a Good Laboratory Pipetting guide offering a extensive information of pipetting and factors influencing to the pipetting results.

Compatible with:

Finntip, ART tip, HLT tip

Description & Specifications

Specifications

Finntip Flex tips: 10; 200; 1000
Multiple
0.2 to 1000μL
Kit 2: Finnpipettes F1 0.2-2μL micro, 2-20μL, 20-200μL and 100-1000μL
Finnpipette™ F1
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit